Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNHIT6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2865650.1ML
Description
ZNHIT6 Polyclonal specifically detects ZNHIT6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZNHIT6 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Protein A purified | |
RUO | |
54680 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
BCD1chromosome 1 open reading frame 181, box C/D snoRNA essential 1 homolog, box C/D snoRNA protein 1, C1orf181, FLJ12602, FLJ20729, FLJ20760, MGC131963, NY-BR-75, Serologically defined breast cancer antigen NY-BR-75, Zinc finger HIT domain-containing protein 6, zinc finger, HIT type 6, zinc finger, HIT-type containing 6 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of human ZNHIT6 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction