Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103795 Supplier Invitrogen™ Supplier No. PA5103795
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNRD1.

This gene encodes a DNA-directed RNA polymerase I subunit. The encoded protein contains two potential zinc-binding motifs and may play a role in regulation of cell proliferation. The encoded protein may be involved in cancer and human immunodeficiency virus progression. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNRD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNRD1
Gene Accession No. Q6MFY5, Q791N7, Q9P1U0
Gene Alias 1110014N07Rik; DNA-directed RNA polymerase I subunit RPA12; HTEX6; HTEX-6; hZR14; nuclear RNA polymerase I small specific subunit; nuclear RNA polymerase I small specific subunit Rpa12 like; RNA polymerase I small specific subunit Rpa12; Rpa12; Tctex6; tctex-6; TEX6; transcription-associated zinc ribbon protein; zinc ribbon domain containing 1; zinc ribbon domain containing, 1; zinc ribbon domain-containing protein 1; ZNRD1; ZR14
Gene Symbols ZNRD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNRD1(Accession Q9P1U0), corresponding to amino acid residues M68-C118.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30834, 361784, 66136
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.