Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ZNRF2 Partial ORF (NP_667339, 146 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Quantity:
10 μg
25 μg
Description
Sequence: LPAHLSPHMFGGFKCPVCSKFVSSDEMDLHLVMCLTKPRITYNEDVLSKDAGECAICLEELQ
Specifications
Specifications
Accession Number | NP_667339 |
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 223082 |
Molecular Weight (g/mol) | 32.56kDa |
Name | ZNRF2 (Human) Recombinant Protein (Q01) |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 10 μg |
Immunogen | LPAHLSPHMFGGFKCPVCSKFVSSDEMDLHLVMCLTKPRITYNEDVLSKDAGECAICLEELQ |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction