Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNRF2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP128697IR
Description
ZNRF2 Polyclonal antibody specifically detects ZNRF2 in Human samples. It is validated for Western BlotSpecifications
ZNRF2 | |
Polyclonal | |
Western Blot | |
EC 6.3.2.-, Protein Ells2, RING finger protein 202, RNF202E3 ubiquitin-protein ligase ZNRF2, zinc and ring finger 2, zinc finger/RING finger 2, Zinc/RING finger protein 2 | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human zinc and ring finger 2 using the numbering given in entry NP_667339.1 (GeneID 223082). | |
0.1 mL | |
Zinc Finger | |
223082 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction