Learn More
ZNRF2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302311AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). ZNRF2 (zinc and ring finger 2), also known as RNF202, is a 242 amino acid peripheral membrane protein that contains one RING-type zinc finger and localizes to the lysosome, as well as the endosome and the cell junction. Expressed at high levels in brain tissue, ZNRF2 is thought to function as an E3 ubiquitin-protein ligase that may be involved in the establishment and maintece of neuronal transmission and plasticity. Upon DNA damage, ZNRF2 is subject to phosphorylation, probably by ATR or ATM.Specifications
ZNRF2 | |
Polyclonal | |
Unconjugated | |
ZNRF2 | |
RNF202 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
223082 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8NHG8 | |
ZNRF2 | |
Between 100 and 150 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.