Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNRF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120417 Supplier Invitrogen™ Supplier No. PA5120417
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549. Immunogen sequence: MHPLGLCNNN DEEDLYEYGW VGVVKLEQPE LDPKPCLTVL GKAKRAVQRG ATAVIFDVSE NPEAIDQLNQ GSEDPLKRPV VYVKGADAIK LMNIVNKQKV A.

The ZNRF3 gene encodes a transmembrane E3 ubiquitin ligase that plays a critical role in regulating the Wnt signaling pathway, which is essential for various biological processes, including embryonic development and cell proliferation. ZNRF3 functions by negatively regulating this pathway through ubiquitination and degradation of Frizzled receptors, thereby suppressing the Wnt signaling. Located on chromosome 22, ZNRF3 acts as a tumor suppressor gene by inhibiting oncogenic Wnt signaling and has been implicated in the formation and progression of several cancers, particularly colorectal cancer. Moreover, ZNRF3 interacts with R-spondin proteins which are known to augment Wnt signaling by targeting ZNRF3 for degradation, thus facilitating cell differentiation and proliferation. Alterations in ZNRF3 expression or mutations can lead to aberrant Wnt signaling, contributing to tumorigenesis. Additionally, ZNRF3's role extends to its participation in lipid metabolism and maintenance of bone homeostasis. It has been studied as a potential therapeutic target due to its central involvement in these regulatory pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNRF3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene ZNRF3
Gene Accession No. Q5SSZ7, Q9ULT6
Gene Alias BK747E2.3; CTA-292E10.6; E3 ubiquitin-protein ligase ZNRF3; Gm1167; KIAA1133; novel C3HC4 type Zinc finger (ring finger); rCG35969-like; RING finger protein 203; RING-type E3 ubiquitin transferase ZNRF3; RNF203; similar to ring finger protein 43; zinc and ring finger 3; zinc/RING finger protein 3; ZNRF3
Gene Symbols ZNRF3
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 407821, 685487, 84133
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.