Learn More
Invitrogen™ ZNRF4 Monoclonal Antibody (1A8)

Description
Peptide Sequence: SSSVDFADLP ALFGVPLAPE GIRGYLMEVK PANACHPIEA PRLGNRSLGS IALIRRYDCT FDL.
Specifications
Specifications
| Antigen | ZNRF4 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1A8 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZNRF4 |
| Gene Accession No. | Q8WWF5 |
| Gene Alias | 1700094M01Rik; E3 ubiquitin-protein ligase ZNRF4; KOX18; nixin; RING finger protein 204; RING-type E3 ubiquitin transferase ZNRF4; RNF204; Sperizin; spermatid specific RING zinc finger 1; spzn; Ssrzf1; zinc and ring finger 4; zinc/RING finger protein 4; ZNF139; ZNF36; Znrf4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.