Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNRF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116283
Description
ZNRF4 Polyclonal Antibody for Western Blot
E3 ubiquitin-protein ligase that acts as a negative regulator of NOD2 signaling by mediating ubiquitination and degradation of RIPK2. Also catalyzes ubiquitination and proteasomal degradation of CANX within the endoplasmic reticulum. Could have a role in spermatogenesis (By similarity).
Specifications
| ZNRF4 | |
| Polyclonal | |
| Unconjugated | |
| ZNRF4 | |
| 1700094M01Rik; E3 ubiquitin-protein ligase ZNRF4; KOX18; nixin; RING finger protein 204; RING-type E3 ubiquitin transferase ZNRF4; RNF204; Sperizin; spermatid specific RING zinc finger 1; spzn; Ssrzf1; zinc and ring finger 4; zinc/RING finger protein 4; ZNF139; ZNF36; Znrf4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 148066 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8WWF5 | |
| ZNRF4 | |
| KLH conjugated synthetic peptide between 111-138 amino acids from the N-terminal region of human ZNRF4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction