Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZnT-8/SLC30A8 Antibody (815039) [Janelia Fluor™ 549], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7936I
Description
ZnT-8/SLC30A8 Monoclonal antibody specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
ZnT-8/SLC30A8 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
815039 | |
Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZNT8ZnT-8 | |
E. coli-derived recombinant human ZnT-8, Lys268-Pro359, Accession # Q8IWU4 | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
169026 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction