Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZO-1 Monoclonal Antibody (R26.4C), eBioscience™, Invitrogen™

Catalog No. p-7218426
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-977-637 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-977-637 Supplier Invitrogen™ Supplier No. 14977637
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MAb R26.4C recognizes the tight junction protein 1, ZO-1 (zona occludens) of rat, mouse, canine, and porcine. ZO-1 is a member of the MAGUK family of tight junction associated scaffolding proteins, each containing a PDZ, SH3 and guanylate kinase-like (GUK) domain. ZO-1 is an integral member of the tight junction complex which forms a barrier to paracellular movement of substances, separating apical and basolateral fluids in relation to the epithelial cell layer. ZO-1 forms complexes with family members ZO-2, ZO-3, as well as occludin, cingulin, claudin-1-8, actin, alpha-catenin, the Ras-effector molecule AF-6, ZONAB, Apg-2, and the junctional adhesion molecules (JAM)-A-C. ZO-1 is thought to anchor the actin cytoskeleton to the tight junction. ZO-1 and ZO-2 have been shown to regulate cell cycle and proliferation with recent evidence highlighting the down-regulation of ZO-1 and ZO-2 with cancer progression.

ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZO-1
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone R26.4C
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TJP1
Gene Accession No. O97758, P39447
Gene Alias DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1
Gene Symbols TJP1
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21872, 292994, 403752
Target Species Mouse, Rat, Canine
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.