Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZO-1 Recombinant Mouse Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ Plus 555, Invitrogen™
Mouse Recombinant Monoclonal Antibody
$167.00 - $323.00
Specifications
Antigen | ZO-1 |
---|---|
Clone | ZO1-1A12 |
Concentration | 1.0 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Recombinant Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
74-000-2MP555
![]() |
Invitrogen™
740002MP555 |
50 μL |
Each for $323.00
|
|
|||||
50-253-9568
![]() |
Invitrogen™
740002MP55520UG |
20 μL |
Each for $167.00
|
|
|||||
Description
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.Specifications
ZO-1 | |
1.0 mg/mL | |
Recombinant Monoclonal | |
Liquid | |
RUO | |
Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.5 | |
DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1 | |
TJP1 | |
IgG1 | |
Protein A/G | |
TJP1 |
ZO1-1A12 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Alexa Fluor Plus 555 | |
Mouse | |
Human | |
Q07157 | |
7082 | |
Human recombinant ZO-1 fusion protein encompassing amino acids 334-634. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title