Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZO-3 Polyclonal Antibody, Abbomax

Catalog No. 502204545
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4545 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4545 Supplier Abbomax Inc Supplier No. 630030
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: A431; HEK293 Cellular location: Membrane, Cellular junction.

ZO-3 Polyclonal antibody specifically detects ZO-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ZO-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TJP3
Gene Accession No. O95049, Q9QXY1
Gene Alias tight junction protein 3; tight junction protein 3 (zona occludens 3); tight junction protein ZO-3; TJP3; TJP-3; ZO3; ZO-3; Zona occludens protein 3; zonula occludens protein 3
Host Species Rabbit
Immunogen A synthetic peptide from 902-919 amino acids of human ZO-3 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27134, 27375, 314640
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.