Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZoBell's Solution, Modified, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical

Click to view available options
Quantity:
120 mL
1 L
Specifications
Specifications
Chemical Name or Material | ZoBell's Solution |
Packaging | Amber Poly Bottle |
Name Note | Oxidation-Reduction Potential (ORP) Standard |
CAS Min % | 97.0, 0.73, 0.72, 7.0E-4 |
CAS Max % | 100.0, 0.76, 0.75, 7.0E-4 |
CAS | 13746-66-2, 14459-95-1, 7447-40-7, 7732-18-5 |
Physical Form | Liquid |
Color | Yellow |
Boiling Point | 100°C |
Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction