Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZoBell's Solution, Oxidation-Reduction Potential (ORP) Standard, Ricca Chemical
SDP

Catalog No. 98801
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Poly Bottle
Black Poly Bottle

Specifications

Chemical Name or Material ZoBell's Solution
Packaging Black Poly Bottle
Name Note Oxidation-Reduction Potential (ORP) Standard
CAS Min % 97.0, 0.73, 0.23, 0.11
CAS Max % 100.0, 0.76, 0.24, 0.11
CAS 13746-66-2, 7447-40-7, 14459-95-1, 7732-18-5
Physical Form Liquid
Boiling Point 100°C
Quantity 4 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.