Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Zoledronic Acid
GREENER_CHOICESDP

Catalog No. 611150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-115-0 50 mg
61-111-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-115-0 Supplier Tocris Bioscience™ Supplier No. 6111/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent farnesyl diphosphate (FPP) synthase inhibitor

Zoledronic Acid is a potent bisphophonate farnesyl diphosphate (FPP) synthase inhibitor (IC50 = 20 nM). Inhibits osteoclast-mediated bone resorption. Also inhibits Ras signaling and tumor growth, and induces apoptosis in pancreatic cancer cells. Reverses epithelial-mesenchymal transition and inhibits breast cancer cell renewal via inactivation of NF-κB.

Specifications

Chemical Name or Material Ras GTPase Inhibitors
CAS 118072-93-8
Synonym [1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid
Quantity 50 mg
Solubility Soluble to 50mM in 1.1eq. NaOH and to 3mM in water
Molecular Formula C5H10N2O7P2
Formula Weight 272.09
Physical Form Solid
Color Off-white
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.