Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZONAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535151
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535151 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535151 Supplier Invitrogen™ Supplier No. PA535151
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with rat based on sequence homology.

Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZONAB
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene YBX3
Gene Accession No. P16989, Q9JKB3
Gene Alias cold shock domain protein A; cold shock domain-containing protein A; cold-shock domain containing A1; cold-shock domain protein A; Csda; CSDA1; Dbpa; DNA-binding protein A; Dpba; MSY3; MSY4; Muscle Y-box protein YB2; oxyR; RYB-A; single-strand DNA-binding protein NF-GMB; Y box binding protein 3; Y box protein 3; Yb2; Y-box binding protein 3; Y-box protein 3; Y-box-binding protein 3; Y-box-binding protein A; Ybx3; ZO-1-associated nucleic acid-binding protein; ZONAB; ZONAB A; ZONAB B
Gene Symbols YBX3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 152-181 amino acids from the central region of human CSDA.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56449, 8531
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.