Learn More
ZONAB Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303070AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.Specifications
ZONAB | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P16989 | |
YBX3 | |
Between 250 and 300 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
YBX3 | |
CSDA1, DBPA, ZONAB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8531 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.