Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Zoniporide dihydrochloride
GREENER_CHOICE

Catalog No. 272710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-271-0 10 mg
27-275-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-271-0 Supplier Tocris Bioscience™ Supplier No. 2727/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective NHE1 inhibitor

Sodium-hydrogen exchanger isoform 1 (NHE1) inhibitor that displays selectivity over other NHE isoforms (Ki values are 14, 2200 and 220000 nM for human NHE1, human NHE2 and rat NHE3 respectively). Inhibits NHE1 dependent 22Na+ uptake in vitro (IC50 = 14 nM) and provides cardioprotection from myocardial ischemic injury in vivo (EC50 = 0.25 nM). Also inhibits MMP2/9 activity and invasion in breast cancer cells.

Chemical Identifiers

CAS 241799-10-0
Molecular Formula C17H18Cl2N6O

Specifications

Quantity 10 mg
Formula Weight 393.27
Purity >99%
Molecular Weight (g/mol) 393.272
Chemical Name or Material Zoniporide dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.