Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZP-2, Rat anti-Mouse, Clone: IE-3, Millipore Sigma™

Catalog No. MABE114225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE114225 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE114225 Supplier MilliporeSigma Supplier No. MABE114225UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-ZP-2, clone IE-3, Cat. No. MABE1142, is a highly specific rat monoclonal antibody, that targets Zona pellucida sperm-binding protein 2 and has been tested in ELISA, Immunoprecipitation, Immunofluorescence and Western Blotting.

Zona pellucida sperm-binding protein 2 (UniProt: P20239; also known as Zona pellucida glycoprotein 2, Zp-2, Zona pellucida protein A) is encoded by the Zp2 (also known as Zp-2, Zpa) gene (Gene ID: 22787) in murine species. The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed of three glycoproteins with various functions during fertilization and preimplantation development. The mammalian zona pellucida mediates species-specific sperm binding, induction of the acrosome reaction, and prevents post-fertilization polyspermy. ZP2 is expressed during the 2-week growth phase of oogenesis, prior to ovulation and may act as a secondary sperm receptor and is proteolytically cleaved in the N-terminal part by the metalloendopeptidase ASTL and is exocytosed from cortical granules after fertilization, yielding an N-terminal peptide of about 30 kDa which remains covalently attached to the C-terminal peptide via disulfide bond(s). This cleavage plays an important role in the post-fertilization block to polyspermy. Female mice lacking Zp2 gene do not form a stable zona matrix and are sterile. (Ref.: Hinsch, KD and Hinsch, E. (1999). Andrologia.31(5):320-322.)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZP-2
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IE-3
Conjugate Unconjugated
Dilution Immunoprecipitation Analysis: A representative lot detected ZP-2 in Immunoprecipitation applications (East, I.J., et. al. (1984). J Cell Biol. 98(3):795-800).Immunofluorescence Analysis: A representative lot detected ZP-2 in Immunofluorescence applications (East, I.J., et. al. (1984). J Cell Biol. 98(3):795-800).Immunofluorescence A representative lot detected ZP-2 in Immunofluorescence applications (Avella, M.A., et. al. (2016). Sci Transl Med. 8(336):336ra60).ELISA Analysis: A representative lot detected ZP-2 in Western Blotting applications (East, I.J., et. al. (1984). J Cell Biol. 98(3):795-800; Sun, W., et. al. (1999). Biol Reprod. 60(4):900-7).Western Blotting Analysis: A representative lot detected ZP-2 in ELISA applications (Sun, W., et. al. (1999). Biol Reprod. 60(4):900-7).
Formulation Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Zona pellucida sperm-binding protein 2;Zona pellucida glycoprotein 2;Zona pellucida protein A
Gene Symbols Zp2;Zp-2;Zpa
Host Species Rat
Immunogen Purified mouse zonae pellucidae.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity Clone IE-3 detects Zona pellucida sperm-binding protein 2 in murine species.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.