Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZP-2 Rat anti-Mouse, Clone: IE-3, MilliporeSigma™

Catalog No. MABE1142MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1142MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1142MI Supplier MilliporeSigma Supplier No. MABE1142
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-ZP-2, clone IE-3 is a highly specific rat monoclonal antibody that targets Zona pellucida sperm-binding protein 2 and has been tested in ELISA, Immunofluorescence, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZP-2
Applications Western Blot, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone IE-3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Zp2,Zp-2,Zpa
Host Species Rat
Immunogen Purified mouse zonae pellucidae.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_035905.1
Test Specificity Clone IE-3 detects Zona pellucida sperm-binding protein 2 in murine species.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.