Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2843690.1ML
Description
ZP1 Polyclonal specifically detects ZP1 in Human samples. It is validated for Western Blot.Specifications
ZP1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
22917 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
KIAA0395TIX1, triple homeobox 1, Triple homeobox protein 1, Zinc finger and homeodomain protein 3, zinc fingers and homeoboxes 3, zinc fingers and homeoboxes protein 3 | |
The immunogen is a synthetic peptide directed towards the middle region of human ZP1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction