Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZP2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297981DL594
Description
ZP2 Polyclonal specifically detects ZP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZP2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Zona pellucida glycoprotein 2, zona pellucida glycoprotein 2 (sperm receptor), zona pellucida glycoprotein ZP2, Zona pellucida protein A, zona pellucida sperm-binding protein 2, Zp-2 | |
Produced in rabbits immunized with E. coli-derived Human ZP2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
7783 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction