Learn More
Invitrogen™ ZP2 Polyclonal Antibody

Description
Immunogen sequence: PNCTYILDPE KLTLRATYDN CTRRVHGGHQ MTIRVMNNSA ALRHGAVMYQ FFCPAMQVEE TQGLSASTIC QKDFMSFSLP RVFSGLADDS KGTKVQMGWS IEVGDGARAK TLTLPEAMKE GFSLLIDNHR MTFHVPFNAT GVT Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 45%.
Specifications
Specifications
| Antigen | ZP2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Zp2 |
| Gene Accession No. | Q05996 |
| Gene Alias | 75-kDa rabbit zona pellucida protein; Processed zona pellucida sperm-binding protein 2; zona pellucida 2 glycoprotein; zona pellucida A protein; zona pellucida glycoprotein 2; zona pellucida glycoprotein 2 (sperm receptor); zona pellucida glycoprotein ZP2; zona pellucida glycoprotein ZPA; zona pellucida protein A; Zona pellucida sperm-binding protein 2; ZP2; Zp-2; ZPA |
| Gene Symbols | Zp2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.