Learn More
Invitrogen™ ZP3 Polyclonal Antibody

Description
Immunogen sequence: LTFSLRLMEE NWNAEKRSPT FHLGDAAHLQ AEIHTGSHVP LRLFVDHCVA TPTPDQNASP YHTIVDFHGC LVDGLTDASS AFKVPRPGPD TLQFTVDVFH FANDSRNMIY ITCHLKVTLA EQDPDELNKA CSFSKPSNSW FPVEGSADIC QCCNKGDCGT PSHSRRQPHV MSQWSRSASR NRRHVTEEAD VTVGPLIFLD RRGDHEVEQW ALPSDTSVVL L; Positive Samples: SKOV3; Cellular Location: Cell membrane, Single-pass type I membrane protein, extracellular matrix, extracellular space.
Specifications
Specifications
| Antigen | ZP3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ZP3 |
| Gene Accession No. | P21754, P97708 |
| Gene Alias | Processed zona pellucida sperm-binding protein 3; Sperm receptor; zona pellucida C; zona pellucida C protein; zona pellucida glycoprotein 3; zona pellucida glycoprotein 3 (sperm receptor); zona pellucida glycoprotein 3 preproprotein; zona pellucida glycoprotein 3A (sperm receptor); zona pellucida glycoprotein 3B; zona pellucida glycoprotein ZP3; zona pellucida glycoprotein ZP3B; zona pellucida protein C; Zona pellucida sperm-binding protein 3; Zp3; Zp-3; ZP3A; ZP3A/ZP3B; ZP3B; Zp-3B; ZPC |
| Gene Symbols | ZP3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.