Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZPBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563603 Supplier Invitrogen™ Supplier No. PA563603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSCEISLLKS ECHRVKMQRA GLQNELFFAF SVSSLDTEKG PKRCTDHNCE PYKRLFKAKN LIERFFNQQV EILG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

ZPBP (also known as ZPBP1), named for its binding to the oocyte zona pellucida, is a spermatid and spermatozoon protein that localizes to the acrosome. ZPBP may be implicated in gamete interaction during fertilization. It has been reported that absence of ZPBP1 prevents acrosome compaction and leads to acrosome fragmentation and sterility in male mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZPBP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Zpbp
Gene Accession No. Q9BS86
Gene Alias 4930486K01Rik; Iam38; Inner acrosomal membrane IAM38; Sp38; zona pellucida binding protein; zona pellucida binding protein 1; zona pellucida-binding protein 1; ZPBP; ZPBP1
Gene Symbols Zpbp
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZPBP. Recombinant protein control fragment (Product #RP-101775).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11055
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.