Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZPBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120564
This item is not returnable.
View return policy
Description
Positive test controls include: Mouse brain. Immunogen sequence: HLVRLPRAFR LTKDSVKIVG STSFPVKAYV MLHQKSPHVL CVTQQLRNAE LIDPSFQWYG PKGKVVSVEN RTAQITSTGS LVFQNFEESM SGIYTCFLEY KPTVEEIVKR LQLKYAIYAY REPHYYYQFT ARYHAAPCNS IYNISFEKKL LQILSK.
ZPBP (also known as ZPBP1), named for its binding to the oocyte zona pellucida, is a spermatid and spermatozoon protein that localizes to the acrosome. ZPBP may be implicated in gamete interaction during fertilization. It has been reported that absence of ZPBP1 prevents acrosome compaction and leads to acrosome fragmentation and sterility in male mice.
Specifications
ZPBP | |
Polyclonal | |
Unconjugated | |
Zpbp | |
4930486K01Rik; Iam38; Inner acrosomal membrane IAM38; Sp38; zona pellucida binding protein; zona pellucida binding protein 1; zona pellucida-binding protein 1; ZPBP; ZPBP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11055, 498415, 53604 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.76 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q62522, Q9BS86 | |
Zpbp | |
Recombinant fusion protein containing a sequence corresponding to amino acids 45-200 of human ZPBP (NP_008940.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction