Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZPK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567220 Supplier Invitrogen™ Supplier No. PA567220
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLHGNTMEKLI KKRNVPQKLS PHSKRPDILK TESLLPKLDA ALSGVGLPGC PKGPPSPGRS RRGKTRHRKA SAKGSCGDL Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

ZPK is a member of serine/threonine protein kinase family. This kinase contains a leucine-zipper domain, and is predominately expressed in neuronal cells. The phosphorylation state of this kinase in synaptic terminals was shown to be regulated by membrane depolarization via calcineurin. This kinase forms heterodimers with leucine zipper containing transcription factors, such as cAMP responsive element binding protein (CREB) and MYC, and thus may play a regulatory role in PKA or retinoic acid induced neuronal differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZPK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP3K12
Gene Accession No. Q12852
Gene Alias DLK; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; leucine-zipper protein kinase; M3KC; MAP3K12; MAPK-upstream kinase; MEKK12; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; PK; protein kinase MUK; zipper (leucine) protein kinase; Zpk; ZPKP1
Gene Symbols MAP3K12
Host Species Rabbit
Immunogen Recombinant Human ZPK. Recombinant protein control fragment (Product #RP-107856).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.