Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZPLD1 Monoclonal Antibody (1D8)
GREENER_CHOICE

Catalog No. PIMA524518
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524518 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524518 Supplier Invitrogen™ Supplier No. MA524518
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: WNVLMDYCYT TPSGNPNDDI RYDLFLSCDK DPQTTVIENG RSQRGRFSF EVFRFVKHKN QKMSTVFLHCV TKLCRADDCP FLMPICSHRE RRDAGRRTTW.

ZPLD1 is a protein coding gene. Glycoprotein which is a component of the gelatinous extracellular matrix in the cupulae of the vestibular organ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZPLD1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZPLD1
Gene Accession No. Q8TCW7
Gene Alias 9430016A21Rik; Cupulin; zona pellucida like domain containing 1; zona pellucida-like domain containing 1; zona pellucida-like domain-containing protein 1; Zona pellucida-like domain-containing protein 1, secreted form; ZP domain-containing protein 1; ZPLD1
Gene Symbols ZPLD1
Host Species Mouse
Immunogen ZPLD1 (NP_778226, 244 a.a. ~ 343 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131368
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.