Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZPR1 Recombinant Rabbit Monoclonal Antibody (JE58-88)
GREENER_CHOICE

Catalog No. PIMA544894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544894 Supplier Invitrogen™ Supplier No. MA544894
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ZPR1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene is found in the cytoplasm of quiescent cells but translocates to the nucleolus in proliferating cells. The encoded protein interacts with survival motor neuron protein (SMN1) to enhance pre-mRNA splicing and to induce neuronal differentiation and axonal growth. Defects in this gene or the SMN1 gene can cause spinal muscular atrophy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Nov 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZPR1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE58-88
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene ZPR1
Gene Accession No. O75312, Q62384
Gene Alias AI303781; fc17g10; hypothetical protein LOC504523; RGD1562173; si:rp71-56i13.4; wu:fc17g10; Zfp259; zgc:56478; Zinc finger protein 259; zinc finger protein ZPR1; ZNF259; Zpr1; ZPR1 zinc finger
Gene Symbols ZPR1
Host Species Rabbit
Immunogen Recombinant protein within human ZPR1 aa 360-459/459.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22687, 8882
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.