Learn More
ZPR9 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304075AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
ZNF622 may behave as an activator of the bound transcription factor, MYBL2, and be involved in embryonic development.Specifications
ZPR9 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q91VY9, Q969S3 | |
Zfp622, ZNF622 | |
Between 25 and 75 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF622 | |
Zinc finger protein 622; Zinc finger-like protein 9; ZNF622; ZPR9; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
52521, 90441 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.