missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZRANB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598975 Supplier Invitrogen™ Supplier No. PA598975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZRANB1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZRANB1 is a positive regulator of the Wnt signaling pathway that specifically cleaves 'Lys-63'-linked ubiquitin chains. It acts by deubiquitinating APC protein, a negative regulator of Wnt-mediated transcription and may also modulate TNF-alpha signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZRANB1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZRANB1
Gene Accession No. Q9UGI0
Gene Alias 9330160G10Rik; D7Wsu87e; DKFZp762P2216; Gm1956; hTrabid; Trabid; TRAF-binding domain-containing protein; TRAF-binding protein; TRAF-binding protein domain; ubiquitin thioesterase ZRANB1; Zinc finger Ran-binding domain-containing protein 1; zinc finger RANBP2-type containing 1; zinc finger, RAN-binding domain containing 1; ZRANB1
Gene Symbols ZRANB1
Host Species Rabbit
Immunogen Recombinant Human Ubiquitin thioesterase ZRANB1 protein (111-397AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54764
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.