Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZRANB2 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10055322FR
Description
ZRANB2 Polyclonal antibody specifically detects ZRANB2 in Human samples. It is validated for Western BlotSpecifications
ZRANB2 | |
Polyclonal | |
Western Blot | |
Oocyte-specific maternal effect factor, zygote arrest 1, zygote arrest protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human zinc-finger protein 265 (zinc-finger, splicing) using the numbering given in entry NP_005446.2 (GeneID 9406). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9406 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction