Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZRANB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565143 Supplier Thermo Scientific Supplier No. PA565143
Add to Cart
Add to Cart

ZRANB3 Polyclonal Antibody

ZRANB3 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZRANB3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZRANB3
Gene Accession No. Q5FWF4
Gene Alias 4933425L19Rik; AH2; AI316834; Annealing helicase 2; C730006D09; DNA annealing helicase and endonuclease ZRANB3; DNA annealing helicase ZRANB3; Endonuclease ZRANB3; RGD1564952; Zinc finger Ran-binding domain-containing protein 3; zinc finger RANBP2-type containing 3; zinc finger, RAN-binding domain containing 3; ZRANB3
Gene Symbols ZRANB3
Host Species Rabbit
Immunogen Recombinant Human ZRANB3. Recombinant protein control fragment (Product #RP-104657).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.