Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZRSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549190
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549190 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549190 Supplier Invitrogen™ Supplier No. PA549190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZRSR1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZRSR1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZRSR2P1
Gene Accession No. Q15695
Gene Alias U2AF1L1; U2AF1P; U2AF1RS1; U2AF1-RS1; U2AFBPL; ZC3H21; zinc finger CCCH-type, RNA binding motif and serine/arginine rich 1; ZRSR1
Gene Symbols ZRSR2P1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 377-403 amino acids from the C-terminal region of human ZRSR1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.