Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZRSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112907
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112907 Supplier Invitrogen™ Supplier No. PA5112907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZRSR2 Polyclonal Antibody for IHC (P), ELISA

This gene encodes an essential splicing factor. The encoded protein associates with the U2 auxiliary factor heterodimer, which is required for the recognition of a functional 3' splice site in pre-mRNA splicing, and may play a role in network interactions during spliceosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZRSR2
Applications ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZRSR2
Gene Accession No. Q15696
Gene Alias 35kDa; 5031411E02Rik; A230052C13Rik; C77286; CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2; Renal carcinoma antigen NY-REN-20; RGD1559763; U2 small nuclear ribonucleoprotein auxiliary factor (U2AF), 35 kDa, related sequence 2; U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2; U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2; U2 small nuclear RNA auxiliary factor 1-like 2; U2(RNU2) small nuclear RNA auxiliary factor 1-like 2; U2AF1L2; U2AF1RS2; U2AF1-RS2; U2AF35-related protein; URP; ZC3H22; zinc finger (CCCH type), RNA binding motif and serine/arginine rich 2; zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2; zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2; ZRSR2
Gene Symbols ZRSR2
Host Species Rabbit
Immunogen Recombinant Human U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-related protein 2 protein (191-372AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8233
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.