Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN16 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP185184
Description
ZSCAN16 Polyclonal specifically detects ZSCAN16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ZSCAN16 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
dJ265C24.3, FLJ22191, zinc finger and SCAN domain containing 16, zinc finger and SCAN domain-containing protein 16, Zinc finger protein 392ZNF392, Zinc finger protein 435ZNF435 | |
Rabbit | |
Affinity Purified | |
RUO | |
80345 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ZSCAN16 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DLSKHRRTHTGEKPYKCDECGKAFIQRSHLIGHHRVHTGVKPYKCKECGKDFSGRTGLIQHQRIHTGEKPYECDECGR | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction