Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN18 Monoclonal Antibody (OTI4B7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505243S


Description
ZSCAN18 Monoclonal antibody specifically detects ZSCAN18 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
ZSCAN18 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TBC5 | |
Mouse | |
Affinity chromatography | |
RUO | |
65982 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunocytochemistry, Western Blot | |
OTI4B7 | |
Unconjugated | |
ZSCAN18 | |
EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18 | |
Full length human recombinant protein of human ZSCAN18 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction