Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN2 Antibody (PCRP-ZSCAN2-1F8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314151PE
This item is not returnable.
View return policy
Description
ZSCAN2 Monoclonal antibody specifically detects ZSCAN2 in Human samples. It is validated for Flow CytometrySpecifications
ZSCAN2 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
54993 | |
Store at 4°C in the dark. | |
Purified |
Flow Cytometry | |
PCRP-ZSCAN2-1F8 | |
Flow Cytometry | |
FLJ20595, zfp-29, ZFP29, zinc finger and SCAN domain containing 2, zinc finger and SCAN domain-containing protein 2, Zinc finger protein 29 homolog, Zinc finger protein 854, ZNF854zinc finger protein 29 | |
Recombinant human fragment (aa 55-135) of ZSCAN 2 protein (Uniprot: Q7Z7L9) | |
0.1 mL | |
Primary | |
Human | |
Antibody | |
IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction