Learn More
Invitrogen™ ZSCAN2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ZSCAN2.
Specifications
Specifications
| Antigen | ZSCAN2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ZSCAN2 |
| Gene Accession No. | Q07230, Q7Z7L9 |
| Gene Alias | Zfp29; Zfp-29; zinc finger and SCAN domain containing 2; zinc finger and SCAN domain-containing protein 2; Zinc finger protein 29; zinc finger protein 29 homolog; Zinc finger protein 854; ZNF854; Zscan2 |
| Gene Symbols | ZSCAN2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.