Learn More
ZSCAN21 Mouse anti-Canine, Human, Rat, Clone: OTI1H7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506150

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.Specifications
ZSCAN21 | |
Monoclonal | |
Unconjugated | |
ZSCAN21 | |
NY-REN-21, ZNF38, Zipro1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
304342, 489848, 7589 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1H7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5A6 | |
ZKSCAN1, Zscan21 | |
Full length human recombit protein of human ZSCAN21 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.