Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN21/ZFP38 Antibody (OTI2E5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274956H
Description
ZSCAN21/ZFP38 Monoclonal specifically detects ZSCAN21/ZFP38 in Human, Mouse, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZSCAN21/ZFP38 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ZSCAN21 | |
Full length recombinant protein of human ZSCAN21/ZFP38 (NP_666019) produced in HEK293T cells. | |
0.1 mL | |
7589.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2E5 | |
Western Blot, Immunohistochemistry | |
DKFZp434L134, KOX25, NY-REN-21, Renal carcinoma antigen NY-REN-21, Zfp-38, ZFP38, zinc finger and SCAN domain containing 21, zinc finger and SCAN domain-containing protein 21, zinc finger protein 38, zinc finger protein 38 (KOX 25), Zinc finger protein 38 homolog, zinc finger protein NY-REN-21 antigen, Zipro1, ZNF38DKFZp686H10254 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction