Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN4 Antibody (OTI1A6), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274957JF549
Description
ZSCAN4 Monoclonal specifically detects ZSCAN4 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZSCAN4 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZSCAN4 | |
Recombinant protein fragment corresponding to amino acids 164-433 of human ZSCAN4 (NP_689890) produced in E.coli. | |
0.1 mL | |
201516.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1A6 | |
Western Blot, Immunohistochemistry | |
FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction