Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN4 Monoclonal Antibody (2-7)
GREENER_CHOICE

Catalog No. PIMA534914
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534914 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534914 Supplier Invitrogen™ Supplier No. MA534914
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293 tranfected Zscan4, 293T, HepG2, human liver tissue.

Zytotic genome activation (ZGA) is the first and most critical transition event from the maternal genetic to the embryonic genetic program and the major burst of ZGA occurs only in the late 2-cell stage. ZSCAN4 encodes a SCAN domain and four zinc finger domains and is expressed in late 2-cell embryos and embryonic stem cells. Loss-of-function studies using siRNA shows delays in the progression from the 2-cell to the 4-cell stage and subsequent embryo development. ZSCAN4 plays an important role in the preimplantation development. Also, ZSCAN4 is involved in telomere maintenance and long term genomic stability in ES cells, suggesting that ZSCAN4 may be of critical importance in maintaining pluripotency in replicating stem cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN4
Applications Western Blot
Classification Monoclonal
Clone 2-7
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene ZSCAN4
Gene Accession No. Q8NAM6
Gene Alias BQ559217; C330039H09Rik; RGD1563625; zinc finger and SCAN domain containing 4; zinc finger and SCAN domain containing 4A; zinc finger and SCAN domain containing 4F; zinc finger and SCAN domain containing protein 4F; Zinc finger and SCAN domain-containing protein 4; zinc finger protein 494; ZNF494; ZSCAN4; Zscan4a; Zscan4f
Gene Symbols ZSCAN4
Host Species Mouse
Immunogen Recombinant protein within Human ZSCAN4 aa 1-138.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201516
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.