Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZSCAN4 Monoclonal Antibody (OTI1A6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526458
Description
ZSCAN4 Monoclonal Antibody for Western Blot, IHC (P)
Zytotic genome activation (ZGA) is the first and most critical transition event from the maternal genetic to the embryonic genetic program and the major burst of ZGA occurs only in the late 2-cell stage. ZSCAN4 encodes a SCAN domain and four zinc finger domains and is expressed in late 2-cell embryos and embryonic stem cells. Loss-of-function studies using siRNA shows delays in the progression from the 2-cell to the 4-cell stage and subsequent embryo development. ZSCAN4 plays an important role in the preimplantation development. Also, ZSCAN4 is involved in telomere maintenance and long term genomic stability in ES cells, suggesting that ZSCAN4 may be of critical importance in maintaining pluripotency in replicating stem cells.
Specifications
ZSCAN4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NAM6 | |
ZSCAN4 | |
Human recombinant protein fragment corresponding to amino acids 164-433 of ZSCAN4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1A6 | |
Unconjugated | |
ZSCAN4 | |
BQ559217; C330039H09Rik; RGD1563625; zinc finger and SCAN domain containing 4; zinc finger and SCAN domain containing 4A; zinc finger and SCAN domain containing 4F; zinc finger and SCAN domain containing protein 4F; Zinc finger and SCAN domain-containing protein 4; zinc finger protein 494; ZNF494; ZSCAN4; Zscan4a; Zscan4f | |
Mouse | |
Affinity chromatography | |
RUO | |
201516 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction