Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZSCAN4C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110610
Description
Immunogen sequence: CSRMFKHARS LSSHQRTHLN KKSELLCVTC QKMFKRVSDR RTHEIIHMPE KPFKCSTCEK SFSHKTNLKS HEMIHTGEMP YVCSLCSRRF RQSSTYHRHL RNYHRSD.
Embryonic stem (ES) cell-specific transcription factor required to regulate ES cell pluripotency. Binds telomeres and plays a key role in genomic stability in ES cells by regulating telomere elongation. Acts as an activator of spontaneous telomere sister chromatid exchange (T-SCE) and telomere elongation in undifferentiated ES cells.
Specifications
ZSCAN4C | |
Polyclonal | |
Unconjugated | |
Zscan4c | |
Gm397; XM_142517; zinc finger and SCAN domain containing 4C; zinc finger and SCAN domain containing protein 4C; Zscan4c; Zscan4d | |
Rabbit | |
Affinity chromatography | |
RUO | |
245109 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.07 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80VJ6 | |
Zscan4c | |
A synthetic peptide corresponding to a sequence within amino acids 400-506 of mouse ZSCAN4 (NP_0010137871). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction