Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541173 Supplier Invitrogen™ Supplier No. PA541173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QDSDIEMAEA PSSVRDDLKD VSSQRASSVN QMRPGEGQAH RELQILPRVP Sequence homology: Human: 100%.

ZSCAN5A may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZSCAN5A
Gene Accession No. Q9BUG6
Gene Alias zinc finger and SCAN domain containing 5; zinc finger and SCAN domain containing 5A; zinc finger and SCAN domain-containing protein 5A; Zinc finger protein 495; ZNF495; ZSCAN5; ZSCAN5A
Gene Symbols ZSCAN5A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ZSCAN5A.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79149
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.