Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZSCAN5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541174
Description
Peptide sequence: KPDASSISQE EPQGEATPVG NRESPGQAGM NSIHSPGPAS PVSHPDGQEA Sequence homology: Human: 100%.
ZSCAN5A may be involved in transcriptional regulation.
Specifications
| ZSCAN5 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN5A | |
| zinc finger and SCAN domain containing 5; zinc finger and SCAN domain containing 5A; zinc finger and SCAN domain-containing protein 5A; Zinc finger protein 495; ZNF495; ZSCAN5; ZSCAN5A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79149 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BUG6 | |
| ZSCAN5A | |
| Synthetic peptide directed towards the middle region of human ZSCAN5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction