Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528011
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528011 Supplier Invitrogen™ Supplier No. PA528011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3. Predicted reactivity: Rhesus Monkey (93%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZSCAN5A may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene ZSCAN5A
Gene Accession No. Q9BUG6
Gene Alias zinc finger and SCAN domain containing 5; zinc finger and SCAN domain containing 5A; zinc finger and SCAN domain-containing protein 5A; Zinc finger protein 495; ZNF495; ZSCAN5; ZSCAN5A
Gene Symbols ZSCAN5A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZSCAN5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.