Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN5C Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549221

Catalog No. PA549221


Only null left
Add to Cart

Description

Description

ZSCAN5C Polyclonal Antibody for Western Blot

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZSCAN5C
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
A6NGD5
ZSCAN5C
KLH conjugated synthetic peptide between 136-162 amino acids from the Central region of human ZSCAN5C.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
ZSCAN5C
ZSCAN5C
Rabbit
Antigen affinity chromatography, Protein A
RUO
649137
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.