Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN5D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP191322
Description
ZSCAN5D Polyclonal specifically detects ZSCAN5D in Human samples. It is validated for Western Blot.Specifications
ZSCAN5D | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ZSCAN5D | |
Synthetic peptide directed towards the middle region of human LOC646698. Peptide sequence RKNLNEHKLIHSGEKPYKCPKCLRAFRRPETLKYHQKTHQETTAPRECEG. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%. | |
Human | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Putative zinc finger and SCAN domain-containing protein 5D, zinc finger and SCAN domain containing 5D, ZNF495D | |
Rabbit | |
Affinity purified | |
RUO | |
646698 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction